To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:oxysterol binding protein-like 9
- Clonality:Polyclonal
- Gene ID:Q96SU4
- Host:Goat
- Reactivity:Human
- Antigen symbol:OSBPL9
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:0.1 mg
- Form:Liquid
- Antigen synonyms:OSBPL9
- Storage buffer:ORP9 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% bovine serum albumin.
- Storage temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:500 ug/mL
- Shipping temperature:4 °C
- Immunogen:ORP9 antibody was raised against a 12 amino acid synthetic peptide near the C-Terminus of ORP9.
- Purification:ORP9 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:10112-802
- Supplier No.:46-112
Specifications
About this item
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 150 kDa and 19 kDa in lysates of human brain, and testis and 55 kDa in lysates of human placenta and Testes after 0.5 ?g/mL antibody staining.5 kDa for NP_683702.1 and NP_683703.1; b: 63.9 kDa for NP_683704.1; c: 70.2 kDa for NP_683705.1; d: 81.8 kDa for NP_683706.1; e: 83.2 kDa for NP_078862.2; f: 84.3 kDa for NP_683707.1). All detected bands were successfully blocked by incubation with the immunizing peptide (and BLAST results with the immunizing peptide sequence did not identify any other proteins to explain the additional bands). Immunohistochemistry: In paraffin embedded human lung shows staining of alveolar macrophages.. Recommended concentration, 2-4 ?g/mL.
ORP9 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: OSBPL9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human