To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:glutamate receptor, ionotropic, N-methyl D-aspartate 1
- Clonality:Polyclonal
- Gene ID:Q05586
- Host:Goat
- Reactivity:Rat
- Antigen symbol:GRIN1
- Conjugation:Unconjugated
- ELISA:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:GRIN1
- Storage buffer:NMDA Receptor antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% bovine serum albumin.
- Storage temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:500 ug/mL
- Shipping temperature:4 °C
- Immunogen:NMDA Receptor antibody was raised against a 12 amino acid synthetic peptide near the internal region of NMDA Receptor.
- Purification:NMDA Receptor antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:10112-030
- Supplier No.:45-971
Specifications
About this item
ELISA: Antibody detection limit dilution 1:1,000. Western Blot: Approximately 110 kDa band observed in rat brain lysates (calculated MW of 105 kDa according to NP_015566.1). Recommended concentration: 2-4 ?g/mL. A minor band of unknown identity was also consistently observed at 23-24 kDa . This band was strong in mouse brain lysates and faint in human brain lysates. The band was successfully blocked by incubation with the immunizing peptide.
NMDA Receptor antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: GRIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat