To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:histamine receptor H1
- Clonality:Polyclonal
- Gene ID:P35367
- Host:Goat
- Reactivity:Human
- Antigen symbol:HRH1
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:0.1 mg
- Form:Liquid
- Antigen synonyms:HRH1
- Storage buffer:Histamine Receptor H1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% bovine serum albumin.
- Storage temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:500 ug/mL
- Shipping temperature:4 °C
- Immunogen:Histamine Receptor H1 antibody was raised against a 14 amino acid synthetic peptide near the C-Terminus of Histamine Receptor H1.
- Purification:Histamine Receptor H1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:10113-324
- Supplier No.:45-731
Specifications
About this item
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 55 kDa and 37 kDa in human duodenum lysate after 0.3 ?g/mL antibody staining.8 kDa according to NP_000852.1. Both detected bands were successfully blocked by incubation with the immunizing peptide (and BLAST results with the immunizing peptide sequence did not identify any other proteins to explain the additional band). Immunohistochemistry: In paraffin embedded human colon shows membranous staining. Recommended concentration, 4-6 ?g/mL.
Histamine Receptor H1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: HRH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human