To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:cytochrome b reductase 1
- Clonality:Polyclonal
- Gene ID:Q53TN4
- Host:Goat
- Reactivity:Human,Mouse
- Antigen symbol:CYBRD1
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:0.1 mg
- Form:Liquid
- Antigen synonyms:CYBRD1
- Storage buffer:Cytochrome B Reductase 1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% bovine serum albumin.
- Storage temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:500 ug/mL
- Shipping temperature:4 °C
- Immunogen:Cytochrome B Reductase 1 antibody was raised against a 15 amino acid synthetic peptide near the C-Terminus of Cytochrome B Reductase 1.
- Purification:Cytochrome B Reductase 1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:10115-852
- Supplier No.:45-452
Specifications
About this item
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: experiments gave bands at approximately 35 kDa and a 26+28 kDa doublet in human colon lysate after 0.1 ?g/mL antibody staining. The detected bands were all successfully blocked by incubation with the immunizing peptide (and BLAST results with the immunizing peptide sequence did not identify any other proteins to explain the additional bands). Immunohistochemistry: In paraffin embedded human testis shows membranous staining in primarily myofibroblasts of the basement membrane surrounding the seminiferous tubules. Recommended concentration, 2-5 ?g/mL.
Cytochrome B Reductase 1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: CYBRD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse