To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:cholecystokinin A receptor
- Clonality:Polyclonal
- Gene ID:O08786
- Host:Goat
- Reactivity:Rat
- Antigen symbol:Cckar
- Conjugation:Unconjugated
- ELISA:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:Cckar
- Storage buffer:CCKA Receptor antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% bovine serum albumin.
- Storage temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:500 ug/mL
- Shipping temperature:4 °C
- Immunogen:CCKA Receptor antibody was raised against a 14 amino acid synthetic peptide near the internal region of CCKA Receptor.
- Purification:CCKA Receptor antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:10115-734
- Supplier No.:45-376
Specifications
About this item
ELISA: Antibody detection limit dilution 1:64,000. Approximately 75 kDa band observed in purified membranes of rat insulinoma cell line RIN14B (calculated MW of 48.4 kDa according to NP_033957.1). Data kindly provided by Dr. Morisset, Sherbrooke, Canada. Data accepted by International Journal of Endocrinology (in press). The observed molecular weight also corresponds to earlier findings in literature with different antibodies (Morisset et al J Histochem Cytochem. 2003 Nov;51 (11) :1501-13; PMID: 14566022). Recommended concentration: 0.1-0.3 ?g/mL.
CCKA Receptor antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: Cckar
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat