To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:cortactin
- Clonality:Polyclonal
- Gene ID:Q14247
- Host:Goat
- Reactivity:Human,Rat,Mouse
- Antigen symbol:CTTN
- Conjugation:Unconjugated
- ELISA:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:CTTN
- Storage buffer:Cortactin antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% bovine serum albumin.
- Storage temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:500 ug/mL
- Shipping temperature:4 °C
- Immunogen:Cortactin antibody was raised against a 12 amino acid synthetic peptide near the N-Terminus of Cortactin.
- Purification:Cortactin antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:10115-370
- Supplier No.:45-044
Specifications
About this item
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 80 kDa band observed in mouse and rat brain lysates (calculated MW of 61.6 kDa according to human NP_005222.2 and of 61.2 kDa according to mouse NP_031829.2). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Paradis et al, J Cell Sci. 2008 Jun 15;121 (Pt 12) :1965-72; PMID 18495841). Recommended concentration: 0.01-0.03 ?g/mL.
Cortactin antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: CTTN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat