To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:histone deacetylase 1
- ChIP:Yes
- Clonality:Polyclonal
- Gene ID:Q13547
- Host:Goat
- Reactivity:Human,Mouse
- Antigen symbol:HDAC1
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoPrecipitation:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:HDAC1
- Storage buffer:HDAC1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% bovine serum albumin.
- Molecular weight:Appox 55 kDa
- Storage temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:500 ug/mL
- Shipping temperature:4 °C
- Immunogen:HDAC1 antibody was raised against a 12 amino acid synthetic peptide near the C-Terminus of HDAC1.
- Purification:Antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:10113-408
- Supplier No.:42-976
Specifications
About this item
Western Blot: Appox 55 kDa band observed in Human Lymph Node and Thymus lysates and approx 70 kDa band observed in lysates of cell lines HeLa and NIH3T3 (calculated MW of 55.1 kDa according to NP_004955.2). The observed 70 kDa band represents the sumoylated protein according to David et al, J Biol Chem. 2002 Jun 28;277(26):23658-63; PMID: 11960997. Recommended concentration: 0.5-2 ?g/mL.
HDAC1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: HDAC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse