To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:tropomyosin 1 (alpha)
- Clonality:Polyclonal
- Gene ID:P09493
- Host:Rabbit
- Isotype:IgG
- Reactivity:Non-human primates,Chicken,Human,Rat,Mouse
- Antigen symbol:TPM1
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:0.05 mg
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:TPM1
- Shipping temperature:RT
- Immunogen:A synthetic phosphopeptide corresponding to the region surrounding S283.
- Purification:Affinity Purified
- Cat. no.:10070-722
- Supplier No.:39-877
Specifications
About this item
Tropomyosin (Tpm-1)consists of a family of highly conserved, widely distributed actin-binding proteins involved in the contractile system of striated and smooth muscles and the cytoskeleton of non-muscle cells. Tropomyosin is composed of two alpha-helical chains arranged as a coiled-coil. It is polymerized end to end along the two grooves of actin filaments and provides stability to the filaments. The encoded protein is one type of alpha helical chain that forms the predominant tropomyosin of striated muscle, where it also functions in association with the troponin complex to regulate the calcium-dependent interaction of actin and myosin during muscle contraction..
ELISA, Western (immuno) blotting, Immunostaining (in testing)
Affinity Purified
Type: Primary
Antigen: TPM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Chicken, Non-Human Primate