To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:chemokine (C-C motif) ligand 26
- Clonality:Monoclonal
- Clone:1.1_3H10-1D11
- Gene ID:Q9Y258
- Host:Mouse
- Neutralisation:Yes
- Reactivity:Human
- Antigen symbol:CCL26
- Conjugation:Unconjugated
- ELISA:Yes
- Size:0.5 mg
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:CCL26
- Shipping temperature:RT
- Immunogen:Eotaxin-3 antibody is produced in BALB/c ICRR F1 mice using highly pure (>98%) recombinant human Eotaxin-3 as the immunizing antigen.
- Purification:This IgG2aK antibody was purified from ascites fluid by Protein A affinity chromatography.
- Cat. no.:10097-932
- Supplier No.:38-238
Specifications
About this item
Neutralization:
To yield one-half maximal inhibition [ND50] of the biological activity of hEotaxin-3 (2.0 ng/mL), a concentration of 1.0-5.0 ?g/mL of this antibody is required.
ELISA:
In a sandwich ELISA (assuming 100 ?L/well), a concentration of 1.0-2.0 ?g/mL of this antibody will detect at least 1000 pg/mL of recombinant human Eotaxin-3 when used with our biotinylated antigen affinity purified anti-human Eotaxin-3 as the detection antibody at a concentration of approximately 0.25-0.50 ?g/mL.
Western Blot:
To detect hEotaxin-3 by Western Blot analysis this antibody can be used at a concentration of 0.25-0.50 ?g/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant hEotaxin-3 is 0.25-0.5 ng/lane, under reducing or non-reducing conditions.
The reconstituted antibody can temporarily be stored at 2-8°C. Long term storage at -20°C or below. Avoid repeated freeze-thaw cycles.
Type: Primary
Antigen: CCL26
Clonality: Monoclonal
Clone: 1.1_3H10-1D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human