To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:chemokine (C-C motif) ligand 23
- Clonality:Monoclonal
- Clone:G5_4E11.3
- Gene ID:P55773
- Host:Mouse
- Reactivity:Human
- Antigen symbol:CCL23
- Conjugation:Unconjugated
- ELISA:Yes
- Size:0.5 mg
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:CCL23
- Shipping temperature:RT
- Immunogen:MIP-3-beta antibody is produced in BALB/c mice using highly pure (>98%) recombinant human MIP-3beta as the immunizing antigen.
- Purification:This IgG1K antibody was purified from cell culture by Protein A affinity chromatography.
- Cat. no.:10097-930
- Supplier No.:38-237
Specifications
About this item
Was product number XP-5714-M
ELISA: In a sandwich ELISA (assuming 100 µL/well), a concentration of 14.0-16.0 µg/mL of this antibody will detect at least 1000 pg/mL of recombinant rat IL-2 when used with our biotinylated antigen affinity purified anti-rat IL-2 (38-205) as the detection antibody at a concentration of at least 2.0-4.0 µg/mL. Western Blot: To detect rat IL-2 by Western Blot analysis this antibody can be used at a concentration of 0.25-0.50 µg/mL. When used in conjunction with compatible secondary reagents the detection limit for recombinant rat IL-2 is 2.0-4.0 ng/lane, under reducing or non-reducing conditions.
This IgG1K antibody was purified from cell culture by Protein A affinity chromatography.
The reconstituted antibody can temporarily be stored at 2-8°C. Long term storage at -20°C or below. Avoid repeated freeze-thaw cycles.
Type: Primary
Antigen: CCL23
Clonality: Monoclonal
Clone: G5_4E11.3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human