To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:collagen, type XVIII, alpha 1
- Clonality:Polyclonal
- Gene ID:P39060
- Host:Rabbit
- Reactivity:Human
- Antigen symbol:COL18A1
- Conjugation:Biotin
- ELISA:Yes
- Size:0.05 mg
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:COL18A1
- Shipping temperature:RT
- Immunogen:Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant hEndostatin (human Endostatin). Human Endostatin specific antibody was purified by affinity chromatography and then biotinylated.
- Cat. no.:10098-372
- Supplier No.:38-195
Specifications
About this item
Endostatin, a cleaved product of the C-terminal domain of collagen XVIII, is an endogenous angiogenesis inhibitor. It was originally found in conditioned media from a murine endothelial tumor cell line, hemangioendothelioma. Endostatin inhibits endothelial cell migration (in vivo and in vitro) and induces endothelial cell apoptosis. It inhibits tumor growth and impairs blood vessel maturation in wound healing. Endostatin has an important role in endothelial cell adhesion and cytoskeletal organization. Endostatin can be found in vessel walls (elastic fibers) and basement membranes.
ELISA:
Direct:
To detect hEndostatin by direct ELISA (using 100 ?L/well antibody solution) a concentration of 0.25 - 1.0 ?g/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with compatible secondary reagents, allows the detection of at least 0.2 - 0.4 ng/well of recombinant hEndostatin.
Sandwich
o detect hEndostatin by sandwich ELISA (using 100 ?L/well antibody solution) a concentration of 0.25 - 1.0 ?g/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with our polyclonal Anti-Human Endostatin as a capture antibody, allows the detection of at least 0.2 - 0.4 ng/well of recombinant hEndostatin.
Western Blot:
To detect hEndostatin by Western Blot analysis this antibody can be used at a concentration of 0.1 - 0.2 ?g/mL. Used in conjunction with compatible secondary reagents the detection limit for recombinant hEndostatin is 1.5 - 3.0 ng/lane, under either reducing or non-reducing conditions.
The reconstituted antibody can temporarily be stored at 2-8°C. Long term storage at -20°C or below. Avoid repeated freeze-thaw cycles.
Type: Primary
Antigen: COL18A1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human