To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Herpes Simplex Virus Antibody [0113] (FITC)
- Clonality:Monoclonal
- Clone:0113
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Virus
- Antigen symbol:HSV
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Size:0.1 mg
- Form:Liquid
- Storage buffer:Herpes Simplex Virus antibody is in a phosphate saline buffer (0.01M, pH 7.2) containing bovine serum albumin (10 mg/ml) as a stabilizer and 0.1% sodium azide preservative.
- Storage temperature:Herpes Simplex Virus antibody can be stored at –20 °C, stable for one year.
- Shipping temperature:4 °C
- Purification:Herpes Simplex Virus antibody is conjugated with high purity isomer I of fluorescein isothiocyanate. Care is taken to ensure complete removal of any free fluorescein from the final product.
- Cat. no.:10070-820
- Supplier No.:35-406
Specifications
About this item
Herpes Simplex Virus antibody is conjugated with high purity isomer I of fluorescein isothiocyanate. Care is taken to ensure complete removal of any free fluorescein from the final product.
Herpes Simplex Virus antibody is in a phosphate saline buffer (0.01M, pH 7.2) containing BSA (10 mg/ml) as a stabilizer and 0.1% sodium azide preservative.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: Herpes Simplex Virus Antibody [0113] (FITC)
Clonality: Monoclonal
Clone: 0113
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: Mouse IgG2b
Reactivity: Virus