To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Clostridium sp. Antibody (HRP)
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Bacteria
- Antigen symbol:Clostridium
- Conjugation:HRP (Horseradish Peroxidase)
- Size:1 mL
- Western blot:Yes
- Form:Liquid
- Storage buffer:Clostridium sp. antibody is in a phosphate saline buffer with bovine serum albumin (10mg/ml) and preserved with 0.002% thimerosal.
- Storage temperature:Clostridium sp. antibody can be stored at –20 °C, stable for one year.
- Shipping temperature:4 °C
- Immunogen:Extract of Cl. perfingens, Cl. sordellii, Cl. novyi, Cl. septicum, Cl. chauvoei
- Purification:Purified IgG fraction of antiserum covalently coupled with highly purified preparation of horseradish peroxidase (RZ>3). Care is taken to insure adequate conjugation while preserving the maximum enzyme activity. Free enzyme is absent.
- Cat. no.:10071-634
- Supplier No.:35-147
Specifications
About this item
Purified IgG fraction of antiserum covalently coupled with highly purified preparation of horseradish peroxidase (RZ>3). Care is taken to insure adequate conjugation while preserving the maximum enzyme activity. Free enzyme is absent.
Clostridium sp. antibody is in a phosphate saline buffer with BSA (10mg/ml) and preserved with 0.002% thimerosal.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: Clostridium sp. Antibody (HRP)
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bacteria