To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:v-yes-1 Yamaguchi sarcoma viral related oncogene homolog
- Clonality:Monoclonal
- Clone:2H8D7
- Gene ID:P07948
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Antigen symbol:LYN
- Conjugation:Unconjugated
- ELISA:Yes
- Size:0.1 mL
- Antigen synonyms:LYN
- Storage buffer:Ascitic fluid containing 0.03% sodium azide.
- Storage temperature:LYN monoclonal antibody can be stored at –20 °C, stable for one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Shipping temperature:4 °C
- Immunogen:Ni-NTA purified truncated recombinant Lyn expressed in E. Coli strain BL21 (DE3).
- Cat. no.:10070-618
- Supplier No.:32-189
Specifications
About this item
Lyn (also known as p53/56 Lyn) is a membrane-associated protein tyrosine kinase (PTK) mostly expressed in hemopoietic cells which is important in cellular signaling. It contains an SH2 and SH3 domain and has been found to be cleaved after activation of caspases in apoptosis. A member of the Src family of PTKs, there are two known isoforms for Lyn which plays an indispensable role in the Fc epsilon RI (Fcer1) and the B-cell IgM receptor signaling pathway and is essential for Syk activation and Lat phosphorylation after Fcer1 aggregation and can also phosphor-ylate Tec on multiple residues. Lyn can also be regulated by IL-2 and IL-3.Lyn is a member of the src family of non-receptor protein tyrosine kinases that is predominantly expressed in haematopoietic tissues. Like all members of the src family, lyn is thought to participate in signal transduction from cell surface receptors that lack intrinsic tyrosine kinase activity. It is associated with a number of cell surface receptors including the B cell antigen receptor and immunoglobulin E receptor (FceRI).
ELISA:Propose dilution 1:10,000.
Determining optimal working dilutions by titration test.
Ascitic fluid containing 0.03% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: LYN
Clonality: Monoclonal
Clone: 2H8D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human