To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:inhibin, alpha
- Clonality:Monoclonal
- Clone:4A2A9 / 4A2F7 / 4A2F2
- Gene ID:P05111
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:INHA
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:0.1 mg
- Western blot:Yes
- Antigen synonyms:INHA
- Storage buffer:Ascitic fluid containing 0.03% sodium azide.
- Storage temperature:INHA(Inhibin alpha) monoclonal antibody can be stored at –20 °C, stable for one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Shipping temperature:4 °C
- Immunogen:Ni-NTA purified truncated recombinant INHA expressed in E. Coli strain BL21 (DE3).
- Purification:Antibody is purified by protein A affinity chromatography.
- Cat. no.:10070-606
- Supplier No.:32-181
Specifications
About this item
INHA(A-inhibin subunit precursor, inhibin alpha subunit ),also called inhibin(alpha) ,which is located on chromosome 2q33-q36. Inhibin is a gonadal protein that preferentially suppresses the secretion of pituitary follicle-stimulating hormone (FSH). Inhibin comprises of two subunits,Inhibin A and B. Inhibin has been shown to regulate gonadal stromal cell proliferation negatively and to have tumor suppressor activity. In addition, serum levels of inhibin have been shown to reflect the size of granulosa cell tumors and can therefore be used as a marker for primary as well as recurrent disease. In addition to their role in endocrine feedback in the reproductive sytem, inhibins subserve local regulatory roles in numerous extragonadal tissues, including brain, adrenal,bone marrow, placenta, and most notably anterior pituitary. Inhibin alpha subunit gene expression is down regulated in human prostate cancer, suggesting a tumor suppressive role.
Western Blot:1:500 - 1:1,000
IHC(P):1:1,000 - 1:2,000
ELISA:Propose dilution 1:10,000
Determining optimal working dilutions by titration test.
Antibody is purified by protein A affinity chromatography.
Purified antibody in PBS containing 0.03% sodium azide
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: INHA
Clonality: Monoclonal
Clone: 4A2A9 / 4A2F7 / 4A2F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human