Order Entry
ContactUsLinkComponent
Anti-ELK1 Mouse Monoclonal Antibody [clone: 3H6D12 / 4H9C8 / 4H9F1]
Anti-ELK1 Mouse Monoclonal Antibody [clone: 3H6D12 / 4H9C8 / 4H9F1]
Catalog #: 10073-234
Supplier:  Prosci
ELK1 Antibody [3H6D12/4H9C8/4H9F1]
Anti-ELK1 Mouse Monoclonal Antibody [clone: 3H6D12 / 4H9C8 / 4H9F1]
Catalog #: 10073-234
Supplier:  Prosci
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ELK1, member of ETS oncogene family
  • Clonality:
    Monoclonal
  • Clone:
    3H6D12 / 4H9C8 / 4H9F1
  • Gene ID:
    P19419
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Antigen symbol:
    ELK1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • ImmunoChemistry:
    Yes
  • Size:
    0.1 mL
  • Western blot:
    Yes
  • Antigen synonyms:
    ELK1
  • Storage buffer:
    Ascitic fluid containing 0.03% sodium azide.
  • Storage temperature:
    ELK1 monoclonal antibody can be stored at –20 °C, stable for one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
  • Shipping temperature:
    4 °C
  • Immunogen:
    Ni-NTA purified truncated recombinant ELK1 expressed in E. Coli strain BL21 (DE3).
  • Cat. no.:
    10073-234
  • Supplier No.:
    32-148

Specifications

About this item

The transcription factor ELK1 is a family of member of ETS oncogene family and of the ternary complex factor (TCF) subfamily,which is located on chromosome Xp11.2 and stimulates transcription. binds to purine-rich DNA sequences. Proteins of the TCF subfamily form a ternary complex by binding to the the serum response factor and the serum reponse element in the promoter of the c-fos proto-oncogene. The protein encoded by this gene is a nuclear target for the ras-raf-MAPK signaling cascade. Elk1 is phosphorylated by MAP kinase pathways at a cluster of S/T motifs at its C terminus,It appears to be a direct target of activated MAP kinase. Biochemical studies indicate that Elk1 is a good substrate for MAP kinase, the kinetics of Elk1phosphorylation and activation correlate with MAP kinase activity, and interfering mutants of MAP kinase block Elk1 activation in vivo. More recent studies have shown that Elk1 is also a target of the Stress Activated Kinase SAPK/JNK. Phosphorylation of Elk1 has also been implicated in synaptic plasticity in the adult hippocampus.

Western Blot:1:500 - 1:1,000
IHC(P):1:500 - 1:1,000
ELISA:Propose dilution 1:10,000.
Determining optimal working dilutions by titration test.

Ascitic fluid containing 0.03% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ELK1
Clonality: Monoclonal
Clone: 3H6D12 / 4H9C8 / 4H9F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Customers Who Bought This Also Bought