Order Entry
ContactUsLinkComponent
Anti-BLK Mouse Monoclonal Antibody [clone: 9D10B7H6 / 9D10A8F8]
Anti-BLK Mouse Monoclonal Antibody [clone: 9D10B7H6 / 9D10A8F8]
Catalog #: 10073-184
Supplier:  Prosci
BLK Antibody [9D10B7H6/9D10A8F8]
Anti-BLK Mouse Monoclonal Antibody [clone: 9D10B7H6 / 9D10A8F8]
Catalog #: 10073-184
Supplier:  Prosci
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    B lymphoid tyrosine kinase
  • Clonality:
    Monoclonal
  • Clone:
    9D10B7H6 / 9D10A8F8
  • Gene ID:
    P51451
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Antigen symbol:
    BLK
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • ImmunoChemistry:
    Yes
  • Size:
    0.1 mL
  • Western blot:
    Yes
  • Antigen synonyms:
    BLK
  • Storage buffer:
    Ascitic fluid containing 0.03% sodium azide.
  • Storage temperature:
    BLK monoclonal antibody can be stored at –20 °C, stable for one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
  • Shipping temperature:
    4 °C
  • Immunogen:
    Ni-NTA purified truncated recombinant BLK expressed in E. Coli strain BL21 (DE3).
  • Cat. no.:
    10073-184
  • Supplier No.:
    32-118

Specifications

About this item

BLK ( B lymphoid tyrosine kinase), with 505-amino acid protein (about 56 kDa), belongs to the Src non-receptor tyrosine kinases family.Different subcellular localizations of Src-family kinases may be important for the regulation of specific cellular processes such as mitogenesis, cytoskeletal organization, and membrane trafficking.Blk is expressed exclusively by B lymphocytes and it is thought to function in a signal transductory pathway specific to this lineage. B lymphoid expression of an active Blk mutant caused proliferation of B progenitor cells and enhanced responsiveness of these cells to interleukin 7. Thus, sustained activation of Blk induces responses normally associated with the pre-BCR.

Western Blot:1:500 - 1:2,000
IHC(P):1:500 - 1:2,000
IHC(F):1:500 - 1:2,000
ELISA:Propose dilution 1:10,000.
Determining optimal working dilutions by titration test.

Ascitic fluid containing 0.03% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: BLK
Clonality: Monoclonal
Clone: 9D10B7H6 / 9D10A8F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Customers Who Bought This Also Bought