To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:SRY (sex determining region Y)-box 2
- Clonality:Polyclonal
- Gene ID:P48431
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Antigen symbol:SOX2
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 ul
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:SOX2
- Storage buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular weight:34 kDa
- Storage temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store SOX2 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human SOX2.
- Purification:Antibody is purified by peptide affinity chromatography method.
- Cat. no.:10111-490
- Supplier No.:31-255
Specifications
About this item
SOX2 is a member of the SRY-related HMG-box (SOX) family of transcription factors involved in the regulation of embryonic development and in the determination of cell fate. The protein may act as a transcriptional activator after forming a protein complex with other proteins. Mutations in this gene have been associated with bilateral anophthalmia, a severe form of structural eye malformation. This intronless gene encodes a member of the SRY-related HMG-box (SOX) family of transcription factors involved in the regulation of embryonic development and in the determination of cell fate. The product of this gene is required for stem-cell maintenance in the central nervous system, and also regulates gene expression in the stomach. Mutations in this gene have been associated with optic nerve hypoplasia and with syndromic microphthalmia, a severe form of structural eye malformation. This gene lies within an intron of another gene called SOX2 overlapping transcript (SOX2OT). Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.
SOX2 antibody can be used for detection of SOX2 by ELISA at 1:12500. SOX2 antibody can be used for detection of SOX2 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: SOX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat