Order Entry
ContactUsLinkComponent
Anti-MMP7 Rabbit Polyclonal Antibody
Anti-MMP7 Rabbit Polyclonal Antibody
Catalog #: 10109-554
Supplier:  Prosci
MMP7 Antibody
Anti-MMP7 Rabbit Polyclonal Antibody
Catalog #: 10109-554
Supplier:  Prosci
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    matrix metallopeptidase 7 (matrilysin, uterine)
  • Clonality:
    Polyclonal
  • Gene ID:
    P09237
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Antigen symbol:
    MMP7
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Size:
    100 ul
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    MMP7
  • Storage buffer:
    Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
  • Molecular weight:
    19 kDa
  • Storage temperature:
    For short periods of storage (days) store at 4 °C. For longer periods of storage, store MMP7 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
  • Shipping temperature:
    4 °C
  • Immunogen:
    Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human MMP7.
  • Purification:
    Antibody is purified by protein A chromatography method.
  • Cat. no.:
    10109-554
  • Supplier No.:
    30-150

Specifications

About this item

Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. MMP7 degrades proteoglycans, fibronectin, elastin and casein and differs from most MMP family members in that it lacks a conserved C-terminal protein domain. The enzyme is involved in wound healing, and studies in mice suggest that it regulates the activity of defensins in intestinal mucosa.Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. The enzyme encoded by this gene degrades proteoglycans, fibronectin, elastin and casein and differs from most MMP family members in that it lacks a conserved C-terminal protein domain. The enzyme is involved in wound healing, and studies in mice suggest that it regulates the activity of defensins in intestinal mucosa. The gene is part of a cluster of MMP genes which localize to chromosome 11q22.3.

MMP7 antibody can be used for detection of MMP7 by ELISA at 1:312500. MMP7 antibody can be used for detection of MMP7 by western blot at 2.5 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MMP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Customers Who Bought This Also Bought