To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Antibody type:Primary
- Antigen name:synaptotagmin binding, cytoplasmic RNA interacting protein
- Antigen symbol:SYNCRIP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Zebrafish,Human,Dog,Rat,Mouse
- Western blot:Yes
- Size:100 ul
- Environmentally Preferable:
- Form:Liquid
- Gene ID:O60506
- Antigen synonyms:SYNCRIP
- Storage buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular weight:69 kDa
- Storage temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store SYNCRIP antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human SYNCRIP.
- Purification:Antibody is purified by protein A chromatography method.
- Cat. no.:10108-100
- Supplier No.:29-374
Heterogenous nuclear ribonucleoprotein (hnRNP) implicated in mRNA processing mechanisms. SYNCRIP may be involved in translationally coupled mRNA turnover. It implicated with other RNA-binding proteins in the cytoplasmic deadenylation/translational and decay interplay of the FOS mRNA mediated by the major coding-region determinant of instability (mCRD) domain. It interacts in vitro preferentially with poly (A) and poly (U) RNA sequences.
SYNCRIP antibody can be used for detection of SYNCRIP by ELISA at 1:312500. SYNCRIP antibody can be used for detection of SYNCRIP by western blot at 1.25 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: SYNCRIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog, Zebrafish