To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:ZPR1 zinc finger
- Clonality:Polyclonal
- Gene ID:O75312
- Host:Rabbit
- Reactivity:Human,Dog,Rat,Mouse
- Antigen symbol:ZNF259
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 ul
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:ZNF259
- Storage buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular weight:51 kDa
- Storage temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store ZNF259 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human ZNF259.
- Purification:Antibody is purified by peptide affinity chromatography method.
- Cat. no.:10107-142
- Supplier No.:28-860
Specifications
About this item
ZNF259 may be a signaling molecule that communicates mitogenic signals from the cytoplasm to the nucleus. ZNF259 binds to the EGFR and is released from the receptor after activation.ZNF259 is essential for cell viability and its interaction with eEF-1alpha contributes to normal cellular proliferation.ZNF259 translocates from the cytoplasm to the nucleus after treatment of cells with mitogens. ZNF259 accumulates in the nucleolus of proliferating cells. Loss of ZNF259 caused disruption of nucleolar function, including preribosomal RNA expression. ZNF259 is an essential protein that is required for normal nucleolar function in proliferating cells.
ZNF259 antibody can be used for detection of ZNF259 by ELISA at 1:312500. ZNF259 antibody can be used for detection of ZNF259 by western blot at 0.03 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: ZNF259
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog