To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:forkhead box N2
- Clonality:Polyclonal
- Gene ID:Q6P4Q2
- Host:Rabbit
- Reactivity:Human,Dog
- Antigen symbol:FOXN2
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 ul
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:FOXN2
- Storage buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular weight:47 kDa
- Storage temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store HTLF antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human HTLF.
- Purification:Antibody is purified by protein A chromatography method.
- Cat. no.:10106-998
- Supplier No.:28-793
Specifications
About this item
Human T-cell leukemia virus enhancer factor (HTLF) may be a forkhead domain binding protein. It may function in the transcriptional regulation of the human T-cell leukemia virus long terminal repeat.
HTLF antibody can be used for detection of HTLF by ELISA at 1:1562500. HTLF antibody can be used for detection of HTLF by western blot at 2.5 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: FOXN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Dog