To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:claudin 23
- Clonality:Polyclonal
- Gene ID:Q6ZW63
- Host:Rabbit
- Reactivity:Human
- Antigen symbol:CLDN23
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 ul
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:CLDN23
- Storage buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular weight:17 kDa
- Storage temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store CLDN23 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human CLDN23.
- Purification:Antibody is purified by peptide affinity chromatography method.
- Cat. no.:10105-688
- Supplier No.:27-911
Specifications
About this item
CLDN23 is a four-transmembrane protein with WWCC motif, defined by W-X (17-22)-W-X (2)-C-X (8-10)-C. It is a candidate tumor suppressor gene implicated in intestinal-type gastric cancer.
CLDN23 antibody can be used for detection of CLDN23 by ELISA at 1:312500. CLDN23 antibody can be used for detection of CLDN23 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: CLDN23
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human