To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:basic helix-loop-helix family, member e22
- Clonality:Polyclonal
- Gene ID:Q8NFJ8
- Host:Rabbit
- Reactivity:Zebrafish,Human,Rat,Mouse
- Antigen symbol:BHLHE22
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 ul
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:BHLHE22
- Storage buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular weight:37 kDa
- Storage temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store BHLHB5 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human BHLHB5.
- Purification:Antibody is purified by protein A chromatography method.
- Cat. no.:10105-238
- Supplier No.:27-594
Specifications
About this item
BHLHB5 is a member of family of basic helix-loop-helix (bHLH) transcription factors. Members of this family have been implicated in many aspects of neural development, including cell growth, differentiation, and cell migration.
BHLHB5 antibody can be used for detection of BHLHB5 by ELISA at 1:62500. BHLHB5 antibody can be used for detection of BHLHB5 by western blot at 2.5 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: BHLHE22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat. Zebrafish