To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:karyopherin alpha 2 (RAG cohort 1, importin alpha 1)
- Clonality:Polyclonal
- Gene ID:P52292
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Antigen symbol:KPNA2
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 ul
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:KPNA2
- Storage buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular weight:58 kDa
- Storage temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store KPNA2 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human KPNA2.
- Purification:Antibody is purified by peptide affinity chromatography method.
- Cat. no.:10104-182
- Supplier No.:26-910
Specifications
About this item
Imported proteins require a nuclear localization sequence (NLS) which generally consists of a short region of basic amino acids or 2 such regions spaced about 10 amino acids apart. Proteins involved in the first step of nuclear import have been identified in different systems. These include the Xenopus protein importin and its yeast homolog, SRP1 (a suppressor of certain temperature-sensitive mutations of RNA polymerase I in Saccharomyces cerevisiae), which bind to the NLS. KPNA2 protein interacts with the NLSs of DNA helicase Q1 and SV40 T antigen and may be involved in the nuclear transport of proteins. KPNA2 also may play a role in V (D)J recombination.The import of proteins into the nucleus is a process that involves at least 2 steps. The first is an energy-independent docking of the protein to the nuclear envelope and the second is an energy-dependent translocation through the nuclear pore complex. Imported proteins require a nuclear localization sequence (NLS) which generally consists of a short region of basic amino acids or 2 such regions spaced about 10 amino acids apart. Proteins involved in the first step of nuclear import have been identified in different systems. These include the Xenopus protein importin and its yeast homolog, SRP1 (a suppressor of certain temperature-sensitive mutations of RNA polymerase I in Saccharomyces cerevisiae), which bind to the NLS. KPNA2 protein interacts with the NLSs of DNA helicase Q1 and SV40 T antigen and may be involved in the nuclear transport of proteins. KPNA2 also may play a role in V (D)J recombination. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.
KPNA2 antibody can be used for detection of KPNA2 by ELISA at 1:62500. KPNA2 antibody can be used for detection of KPNA2 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: KPNA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat