To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:FEZ family zinc finger 1
- Clonality:Polyclonal
- Gene ID:A0PJY2
- Host:Rabbit
- Reactivity:Human
- Antigen symbol:FEZF1
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 ul
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:FEZF1
- Storage buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular weight:52 kDa
- Storage temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store FEZF1 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human FEZF1.
- Purification:Antibody is purified by peptide affinity chromatography method.
- Cat. no.:10103-778
- Supplier No.:26-524
Specifications
About this item
FEZF1 is a transcription repressor. It is involved in the axonal projection and proper termination of olfactory sensory neurons (OSN). It plays a role in rostro-caudal patterning of the diencephalon and in prethalamic formation. Expression of the FEZF1 is required in OSN to cell-autonomously regulate OSN axon projections. FEZF1 regulates non-cell-autonomously the layer formation of the olfactory bulb development and the interneurons. It may be required for correct rostral migration of the interneuron progenitors.
FEZF1 antibody can be used for detection of FEZF1 by ELISA at 1:62500. FEZF1 antibody can be used for detection of FEZF1 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: FEZF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human