- Antibody type:Primary
- Antigen name:zinc finger protein 335
- Antigen symbol:ZNF335
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Size:100 ul
- Form:Liquid
- Gene ID:Q9H4Z2
- Antigen synonyms:ZNF335
- Storage buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular weight:145 kDa
- Storage temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store ZNF335 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human ZNF335.
- Purification:Antibody is purified by peptide affinity chromatography method.
- Cat. no.:10102-186
- Supplier No.:25-529
ZNF335 enhances transcriptional activation by ligand-bound nuclear hormone receptors. However, it does this not by direct interaction with the receptor, but by direct interaction with the nuclear hormone receptor transcriptional coactivator NRC. ZNF335 may function by altering local chromatin structure.The protein encoded by this gene enhances transcriptional activation by ligand-bound nuclear hormone receptors. However, it does this not by direct interaction with the receptor, but by direct interaction with the nuclear hormone receptor transcriptional coactivator NRC. The encoded protein may function by altering local chromatin structure.
ZNF335 antibody can be used for detection of ZNF335 by ELISA at 1:62500. ZNF335 antibody can be used for detection of ZNF335 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: ZNF335
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human