pH Electrodes, IDS (Intelligent Digital Sensors), for Multi, WTW
Supplier: WTW
Certificates
About this item
High quality, enhanced sensor technology combined with most modern measurement electronics.
- IDS electrodes include their serial number and their calibration records
- Measurement integrity benchmarking of the cells by QSC (Quality Sensor Control)
- Ø×L: 12×120 mm
Ordering information: Various cable lengths available.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.