Nunclon™ Sphera™ Microplates, Dishes, and Flasks, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
The Thermo Scientific™ Nunclon™ Sphera™ culture surface allows cells to grow in suspension with virtually no cell attachment
- Optimises spheroid formation
- Supports the consistent formation of cancer spheroids that simulate the 3D structures of tumour growth
- Promotes formation of embryoid bodies (EB)
The Nunclon™ Sphera™ surface minimizes variability and supports the consistent formation of cancer spheroids that simulate the 3D structures of tumor growth, providing a better model system for studying tumor cell progression and efficacy of anticancer agents in vitro.
The new surface supports many different cell types and their ability to generate spheroids; studies show that over time the spheroids grow in volume, indicating that the new surface enables consistent, repeatable cell growth.
Related Information
Certificates
Search by Lot Number10028-030_211901 10028-032_221802 10028-038_421903
Conformity declaration
Customers Who Bought This Also Bought