Stir Bar, Beaker, PTFE, Dynalon
Supplier: Dynalon
Certificates
About this item
The beaker bar fits inside a beaker to create a deep vortex, even at low speeds
Powerful permanent Alnico V magnetic cores. PTFE offers excellent chemical resistance. Always store in pairs to maintain magnetic strength.
Customers Who Bought This Also Bought