Stir Bar Kit, Star-Shaped, PTFE, Dynalon
Catalog #: 30620-476
Supplier: Dynalon
Specifications
- Coating:PTFE
- Description:Star-Shaped Stir Bar Kit
- Cat. no.:30620-476
- Shape:Star
- Weight:0.17800 lbs.
- Supplier No.:304455
Specifications
About this item
Set of 5 stir bars with star-shape
Diameters included are 9.5, 19, 25, 32, and 38mm. Excellent mixing at slow speeds for biologiclal applications. Features larger vortex for more complete mixing. Powerful permanent Alnico V magnetic cores. PTFE offers excellent chemical resistance. Always store in pairs to maintain magnetic strength.
Customers Who Bought This Also Bought