To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Clonality:Polyclonal
- Host:Goat
- Reactivity:Rat
- Antigen symbol:IgG
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:10 mg
- Western blot:Yes
- Epitope:Heavy and Light chain
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,4517577
- Storage temperature:–20 °C
- Concentration:6.29 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Wet Ice
- Immunogen:Rat IgG whole molecule
- Purification:purified by immunoaffinity chromatography
- Cat. no.:RL612-101-033
- Supplier No.:612-101-033
Specifications
About this item
Recommended Dilutions: ELISA: 1:20000 - 1:110000; Western Blot: 1:10000 - 1:40000; Immunochemistry: 1:5000 - 1:20000;
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Rat