To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Tag
- Antigen name:Beta Galactosidase
- Clonality:Polyclonal
- Host:Rabbit
- Antigen symbol:LACZ
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:100 µg
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide
- Storage temperature:–20 °C
- Concentration:1.0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ambient
- Immunogen:Full length native Beta Galactosidase isolated from E.coli
- Purification:purified by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer
- Cat. no.:CARL2004136100
- Supplier No.:200-4136-0100
Specifications
About this item
Beta-Gal Antibody is suitable for immunoblotting (western or dot blot), ELISA, immunofluorescence microscopy, immunoprecipitation, conjugation and most immunological methods requiring high titer and specificity. The antibody recognizes both frozen tissue.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:5000 - 1:10000; Immunochemistry: 1:1500; Other Dilution: User Optimized
Type: Tag
Antigen: lacZ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype:
Reactivity: