To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Control
- Antigen name:IgG2c, k Isotype Ctrl
- Clonality:Monoclonal
- Clone:RTK4174
- Host:Rat
- Isotype:IgG2c kappa
- Reactivity:Rat
- Antigen symbol:IgG2c, k Isotype Ctrl
- Conjugation:Alexa Fluor® 488
- Flow cytometry:Yes
- Size:25 µg
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Storage temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Concentration:0.50 mg/mL
- Shipping temperature:Ambient RT
- Immunogen:Trinitrophenol + KLH
- Purification:The immunoglobulin was purified by affinity chromatography, and conjugated with Alexa Fluor® 488 under optimal conditions.
- Cat. no.:CA400715-BL
- Supplier No.:400715
Specifications
About this item
Alexa Fluor® 488 Rat IgG2c, κ Isotype Ctrl [RTK4174]; Isotype: Rat IgG2c, κ; Reactivity: Rat; Apps: FC, ICFC; Size: 25 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis as negative control. Use at concentrations comparable to those of the specific antibody of interest.
Type: Control
Antigen: IgG2c, k Isotype Ctrl
Clonality: Monoclonal
Clone: RTK4174
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: Rat IgG2c, κ
Reactivity: Rat