Plastic Bag Ties, 4000 Foot Spool, Dynalon
Catalog #: 71070-260
Supplier: Dynalon
Specifications
- Cat. no.:71070-260
- Weight:4.4 lbs.
- Supplier No.:626814
Specifications
About this item
Plastic coated bag ties are extruded of bright red LDPE bonded to an annealed steel wire.
Can be cut to any size.
Customers Who Bought This Also Bought