Heating Cooling Dry Bath Incubator, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Maximize lab space and flexibility with the easy to use, compact, programmable Thermo Scientific™ Digital Heating Cooling Drybath.
- Safety Device: Over temperature protection, Over current detection
- Control Panel: VFD (Vacuum Fluorescent Display), 4 Touch Keys, Dial Knob
- External Material: PP, PC, Powder Coated Steel
- Block Material: Black Anodized Aluminum
- Programs: 10 memories, 10 steps/memory
Wide range of blocks to accommodate almost any size tube required. PID controlled precise temperature setting to the 0.1 degree with a temperature range of 4-95C. Up to 10 programs allowed for memory storage and each program can be set up to 10 steps. Safety features include timer, overheat cut off and water-proof a structure that minimizes influx of reagents or solutions.
Output: Peltier: 160W (max.); Heater: 200W (max.)
Certifications: CE.
Ordering information: Capacity is one block; sold separately.
Specifications
- Catalog No:
- 10054-222
- Temperature uniformity:
- ±0.55 °C (±0.99 °F), at 37 °C
- Temperature stability:
- ±0.15 °C (±0.27 °F), at 37 °C
- Temperature range:
- Ambient –20…95 °C
- Int. W×D×H:
- 99 x 77.5 x 36 mm (3.9 x 3.1 x 1.4")
- Ext. W×D×H:
- 249 x 330 x 168 mm (9.8 x 13 x 6.6")
- Timer:
- 1 min. to 99 hr. 59 min.
Specifications
Customers Who Bought This Also Bought