Rubber Sleeve, Plug-Type Stoppers, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Hollow, plug-type stopper fits into the neck of the apparatus
Sleeve extension fits over the neck for a secure seal. Diaphragm can be punctured with a syringe needle. Use for outgassing NMR tubes, pressure venting or adding reactants via syringe.