Funnel, High Density Polyethylene, Dynalon
Supplier: Dynalon
Certificates
About this item
Funnels are ideal for many applications in the laboratory and industrial markets
Upper portion of funnel is straight sided to helps prevent spill-over. Maximum temperature for HDPE is 110°C.
Customers Who Bought This Also Bought