Cryo-Apron® Aprons, Tempshield
Supplier: Tempshield
Certificates
About this item
These aprons have adjustable straps at waist and neck.
- Protects against cryogenic atmospheres and splashes
- Light and comfortable, reversible with two 100% waterproof liners
- Static free
- Machine washable and dryable
Can be used in cryogenic atmospheres (LN2 vapor phase) and in ultra low temp freezers and blast freezers. Apron is manufactured from nylon which is resistant to most acids and chemicals (do not immerse in liquid nitrogen), and has two layers of semi-porous membrane which is breathable, waterproof, lightweight, non-asbestos, and provides warmth without bulk.
Specifications
- Catalog No:
- 89511-882
- 89511-884
- 89511-881
- 89511-879
- Garment application:
- Food, Laboratory
- Food, Laboratory
- Food, Laboratory
- Food, Laboratory
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Fabric:
- Nylon
- Nylon
- Nylon
- Nylon