Nunc™ Petri Dishes, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Nunc™ Petri Dishes are available in a variety of styles for laboratory use or for use with automatic filling equipment. Most dishes are vented to permit gas exchange during incubation. Bottom flatness is carefully controlled to ensure even distribution of media and organisms. The Bio-Assay dish was developed for use in agar diffusion assays (e.g., antibiotic activity determinations).
- Large square area is especially useful for screening libraries and isolating clones
- Dishes are suitable for culturing of fungi, bacteria and other microorganisms
- Compatible with automated robotic instrumentation
Related Information
Certificates
Search by Lot Number25384-002_161482 25384-002_161944 73521-370_160722 73521-370_162164
Conformity declaration
Thermo Scientific Nunc Cell Culture Plastics Guide
Product Literature
Customers Who Bought This Also Bought