Specifications
- Description:PermaBlue Plus/AP
- Size:30 ml
- Cat. no.:CA10064-752
Specifications
About this item
PermaBlue Plus/AP is a substrate-chromogen system designed to be used for either IHC or ISH when utilizing alkaline phosphatase
It has increase stability of the working solution and produces a distinct bright blue color. It is insoluble in alcohol and xylene substitutes.