About this item
Chemically resistant and non-contaminating, Versilon™ SE-200 chemical transfer tubing provides the outstanding clarity and flexibility of Tygon® tubing combined with an inert fluoropolymer liner.
Versilon™ SE-200 chemical transfer tubing offers crystal clarity and improved flexibility when compared to rigid fluropolymer tubings. Chemically resistant and inert, with a non-contamination fluid path.
Contains no BPA or phthalates. Non-Toxic, taste-free, and odor-free. Versilon™ SE-200 meets FDA 21 CFR Part 177.1550 Criteria for contact with food products.
This tubing formulation is now phthalate-free. Saint-Gobain is proud to be among the first companies to offer sustainable flexible tubing products. The bio-based Versilon™ line combines the high performance standards customers demand with an eco-friendly tubing design. Without sacrificing the flexibility, glass-like clarity, and outstanding bend radius for which Tygon® tubing is known, Versilon™ can handle many applications where flexible tubing of the past could not be used. Its FEP inner liner provides the ultimate in chemical resistance and can accommodate a wide variety of fluids, from corrosives to MEK-based solvents. The inert liner limits the potential of fluid contamination during transfer. Versilon™ SE-200 tubing will not impart odor or taste, making it well suited for food and beverage use.
Specifications
- Catalog No:
- 89404-396
- 89404-388
- 89404-386
- 89404-398
- 89404-394
- 89404-390
- 63010-224
- Model:
- 89404-396
- 89404-388
- 89404-386
- 89404-398
- 89404-394
- 89404-390
- 63010-224
- Bend radius:
- 4"
- 1"
- 1/2"
- 4 1/2"
- 3 1/2"
- 1 1/2"
- 1½"
- Durometer hardness:
- Shore A, 66
- Shore A, 66
- Shore A, 66
- Shore A, 66
- Shore A, 66
- Shore A, 66
- 66 Shore A
- Working pressure:
- 45.0 psi
- 85.0 psi
- 100.0 psi
- 30.0 psi
- 50.0 psi
- 75.0 psi
- 55 psi
Specifications
Related Information
Certificates
Search by Lot Number63010-220_23716504 63010-220_24109755 63010-224_ 22574430 63010-224_24207580 63010-224_24330726