ProSwift RP-1S Protein Columns, Thermo Scientific
Catalog #: 10044-552
Supplier: Thermo Scientific
Specifications
- Column Format:Analytical
- I.D.:4.60 mm
- Length:50 mm
- pH range:1 to 14
- Description:10044-552
- Bed Weight:5.5 mg/ml
- Cat. no.:10044-552
- Packing Material:Insulin
- Phase:Polystyrene divinylbenzene
- Supplier No.:064297
Specifications
About this item
Run protein separations at exceptionally high flow rates with Thermo Scientific™ ProSwift RP columns.
These reversed-phase monolith columns deliver the outstanding resolving power of nonporous analytical media combined with faster separations and analysis than other bead-based columns. ProSwift polymer monolith columns achieve long column lifetime, exhibit reproducibility even after hundreds of runs, and have broad pH stability, providing high stringent wash capability (1 M NaOH).
Customers Who Bought This Also Bought