DNAPac™ PA200 Nucleic Acid Columns, Thermo Scientific
Supplier: Thermo Scientific
Certificates
About this item
For ultra-high-resolution HPLC analysis of synthetic oligonucleotides, use Thermo Scientific™ DNAPac PA200 columns.
These small particle size, strong anion-exchange columns resolve full length oligonucleotides from n–1, n+1, and other failure sequences not possible with other columns. Control the retention times and selectivity by choice of salt, pH, and solvent, and tailor the separation to the requirements of your different oligonucleotide analysis challenges.
Specifications
- Catalog No:
- 10044-492
- 10044-494
- 10044-474
- 10044-476
- 10044-488
- 10044-490
- 10044-350
- 10044-352
- Description:
- 10044-492
- 10044-494
- 10044-474
- 10044-476
- 10044-488
- 10044-490
- 10044-350
- 10044-352
- pH range:
- 2 to 12.5
- 2 to 12.5
- 2 to 12.5
- 2 to 12.5
- 2 to 12.5
- 2 to 12.5
- 2 to 12.5
- 2 to 12.5