Acclaim™ SEC LC Columns, Thermo Scientific
Supplier: Thermo Scientific
Certificates
About this item
Separate water-soluble polymers and oligomers with Thermo Scientific™ Acclaim™ SEC LC Columns.
These resin-based, high-performance, size-exclusion chromatography columns come in two sizes to cover a broad molecular weight range: 5 µm, 300Å for the SEC-300 column and 7 µm, 1000Å for the SEC-1000 column. The combination of a proprietary mono-dispersed multi-pore hydrophilic resin and small particle size provides high-resolution and good linearity of the calibration curve with no inflection points for accurate, reliable analysis of a variety of water-soluble polymers.
Specifications
- Catalog No:
- 10044-882
- 10044-884
- 10044-886
- 10044-918
- 10044-876
- 10044-920
- 10044-878
- 10044-880
- Description:
- 10044-882
- 10044-884
- 10044-886
- 10044-918
- 10044-876
- 10044-920
- 10044-878
- 10044-880
- pH range:
- 2 to 12
- 2 to 12
- 2 to 12
- 2 to 12
- 2 to 12
- 2 to 12
- 2 to 12
- 2 to 12
- Surface area:
- 1000 m²/g
- 1000 m²/g
- 1000 m²/g
- 1000 m²/g
- 1000 m²/g
- 1000 m²/g
- 1000 m²/g
- 1000 m²/g