Siltek™ Cross Inlet Base Seals for Agilent GCs, Thermo Scientific
Catalog #: 10055-776
Supplier: Thermo Scientific
Specifications
- Description:For Single Column, 0.8 mm I.D.
- Cat. no.:10055-776
- Supplier No.:290GA094
Specifications
About this item
Base seals feature Siltek™ coating for a surface with exceptional inertness suitable for the analysis of highly active compounds.
Customers Who Bought This Also Bought