Shelton Scientific IBI® Horizontal Gel Systems: Quick Screening Design, 7×10 cm
Catalog #: 490007-694
Supplier: IBI Scientific
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Quick Screening Horizontal Gel System
- Cat. no.:490007-694
- Supplier No.:IB51000
Specifications
About this item
All models include buffer chamber, safety cover, power cords, casting system with tray, two combs, leveling bubble, and manual.
- 7 x 10cm model designed for quick screening and rapid, linear separation
- of nucleic acids in procedures that do not require buffer recirculation. Buffer chamber Safety cover and power cords
- Casting system with tray
- Two 1.5mm thick 8-tooth combs
- Leveling bubble and manual
Customers Who Bought This Also Bought