ContactUsLinkComponent
Cimarec™ Basic Hotplate Stirrers, Thermo Scientific
Cimarec™ Basic Hotplate Stirrers, Thermo Scientific
Certificates
Cimarec™ Basic Hotplate Stirrers, Thermo Scientific
Certificates
Specifications
  • Catalog No:
  • 97027-352
  • 97027-358
  • Model:
  • 97027-352
  • 97027-358
  • Plate material:
  • Ceramic
  • Ceramic
  • Overall dimensions:
  • 108L×145W×127H mm (7.9×5.7×5"), Surface area: 10.8 cm²
  • 184L×211W×127H mm (11.1×8.3×5"), Surface area:18.4 cm²
  • Electrical:
  • 120V 60 Hz, 405 W
  • 120V 60 Hz, 1086 W
  • Speed range:
  • 100 - 2500 rpm
  • 100 - 2500 rpm
  • Temperature range:
  • 150…538 °C
  • 150…538 °C