About this item
Designed to ensure a bacterial-free fluid path in a wide variety of food processing applications, Tygon S3™ B-44-4X tubing from Saint-Gobain Performance Plastics is now phthalate-free.
Tygon S3™ B-44-4X Food, Milk, and Dairy Tubing features a smooth, non-porous bore that will not trap particulates or promote bacterial growth. Compatible with foods containing a high oil content. Resistant to harsh alkaline cleaners and sanitizers. Excellent alternative to rigid piping systems.
The bio-based Tygon S3™ line combines the high performance standards customers demand with an eco-friendly tubing design. Producers of food, milk, and dairy products insist upon Tygon S3™ B-44-4X for dependable performance in countless filling, draining, transfer, and processing applications. The smooth, non-porous bore inhibits particle entrapment, promoting a sanitary fluid path by minimizing potential for bacteria growth. It offers outstanding resistance to harsh alkaline cleaners and is equally unaffected by commonly used sanitizers. Lightweight and flexible, Tygon S3™ B-44-4X tubing can be put into service quickly. It readily bends to accommodate abrupt corners and obstructions, requiring a minimum of couplings and fittings. Its flexibility can save up to one-third the footage and much of the labor required to install rigid stainless steel or plastic piping. Tygon S3™ B-44-4X tubing is available in large bore sizes (up to 6" inside diameter), making it a flexible alternative to rigid piping systems. It provides complete clarity for positive visual inspection and flow control. A special construction of Tygon S3™ B-44-4X tubing is also available to meet applications involving elevated pressure. In these instances, select Tygon S3™ Pressure Tubing, B-44-4X I.B., which has the identical product features found in Tygon S3™ B-44-4X, with reinforcement added to withstand up to four times the pressure.
Specifications
- Catalog No:
- 89404-224
- 89404-194
- 89404-158
- 89404-156
- 89404-164
- 89404-208
- 89404-162
- 89404-172
- 89404-170
- 89404-168
- 89404-218
- 89404-178
- 89404-176
- 89404-174
- 89404-222
- 89404-188
- 89404-184
- 89404-192
- 89404-190
- Model:
- 89404-224
- 89404-194
- 89404-158
- 89404-156
- 89404-164
- 89404-208
- 89404-162
- 89404-172
- 89404-170
- 89404-168
- 89404-218
- 89404-178
- 89404-176
- 89404-174
- 89404-222
- 89404-188
- 89404-184
- 89404-192
- 89404-190
- Durometer hardness:
- Shore A, 66
- Shore A, 66
- Shore A, 66
- Shore A, 66
- Shore A, 66
- Shore A, 66
- Shore A, 66
- Shore A, 66
- Shore A, 66
- Shore A, 66
- Shore A, 66
- Shore A, 66
- Shore A, 66
- Shore A, 66
- Shore A, 66
- Shore A, 66
- Shore A, 66
- Shore A, 66
- Shore A, 66
- Max. vacuum:
- 25.0"
- 6.0"
- 29.9"
- 29.9"
- 25.0"
- 11.0"
- 29.9"
- 11.0"
- 29.9"
- 16.0"
- 6.0"
- 29.9"
- 25.0"
- 29.9"
- 14.0"
- 11.0"
- 29.9"
- 29.9"
- 25.0"
- Working pressure:
- 34.0 psi
- 19.0 psi
- 60.0 psi
- 100.0 psi
- 34.0 psi
- 25.0 psi
- 43.0 psi
- 25.0 psi
- 50.0 psi
- 28.0 psi
- 20.0 psi
- 47.0 psi
- 34.0 psi
- 43.0 psi
- 27.0 psi
- 25.0 psi
- 40.0 psi
- 44.0 psi
- 34.0 psi