About this item
Rigidly controlled manufacturing processes ensure that Tygothane® Precision Polyurethane Tubing the flexible polyurethane tubing has consistently tight tolerances from lot to lot
Versilon™ C-210-A Multipurpose fuel and oil transfer tubing provides consistently tight dimensional tolerances, excellent abrasion and tear resistance, and excellent resistance to oils, greases, and fuels. It retains flexibility in sub-zero environments.
Made of tough ester-based polyurethane, Versilon™ C-210-A Multipurpose fuel and oil transfer tubing's clarity, high tear strength, and excellent abrasion resistance make it ideal for applications including fuel and lubricant lines, pneumatic lines, abrasive product transfer, and cable jacketing. It also offers exceptional resistance to oils, greases, fuels, and many chemicals. Able to withstand rugged daily use, Versilon™ C-210-A Multipurpose fuel and oil transfer tubing resists weathering and can be safely used in temperatures ranging from –73 to +93 °C (–100 to +200 °F).
While many rubber and plastics materials exhibit resistance to certain solvents, oils, and chemicals, Versilon™ tubing will resist a much wider range of substances. Plasticizer extraction leading to embrittlement is one of the most frequent causes of failure when flexible tubing is exposed to harsh chemicals. Versilon™ Multipurpose fuel and oil transfer tubing is plasticizer-free and remains flexible even when cycled through temperature extremes. The tubing's precision tolerances and high elasticity provide the user with an easy, secure, and worry-free attachment to fittings.
Specifications
- Catalog No:
- 63011-493
- 63011-451
- 63011-489
- 63011-455
- 63011-457
- Model:
- 63011-493
- 63011-451
- 63011-489
- 63011-455
- 63011-457
- Durometer hardness:
- Shore A, 82
- Shore A, 82
- Shore A, 82
- Shore A, 82
- Shore A, 82
- Max. pressure:
- 28.0 psi
- 56.0 psi
- 33.0 psi
- 28.0 psi
- 42.0 psi
- Max. vacuum:
- 15.0"
- 29.9"
- 29.9"
- 20.0"
- 29.9"